Product Details
- SNP ID
-
rs28568876
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:42512107 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGAAAACAATTTGTATGATTTTTA[A/T]TTTTTAGGTAATTATTAATATTACT
- Phenotype
-
MIM: 602534
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SNAP23
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116408563] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SNAP23
- Gene Name
- synaptosome associated protein 23
View Full Product Details