Product Details

SNP ID
rs28464256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43339195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTCATGATGGCATGGAGGCTGC[A/G]GTATCATGCTCCTTGGAGGAGGCAG
Phenotype
MIM: 611246
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAL PubMed Links

Gene Details

Gene
ADAL
Gene Name
adenosine deaminase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012969.3 Intron NP_001012987.1
NM_001159280.2 Intron NP_001152752.1
NM_001324364.1 Intron NP_001311293.1
NM_001324365.1 Intron NP_001311294.1
NM_001324366.1 Intron NP_001311295.1
NM_001324367.1 Intron NP_001311296.1
NM_001324368.1 Intron NP_001311297.1
XM_011521291.2 Intron XP_011519593.1
XM_017021964.1 Intron XP_016877453.1
XM_017021965.1 Intron XP_016877454.1
XM_017021966.1 Intron XP_016877455.1
XM_017021967.1 Intron XP_016877456.1
XM_017021968.1 Intron XP_016877457.1
XM_017021969.1 Intron XP_016877458.1
Gene
LCMT2
Gene Name
leucine carboxyl methyltransferase 2
There are no transcripts associated with this gene.

View Full Product Details