Product Details

SNP ID
rs28613896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43828687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCCTTTCTCACCTTTACATGTT[C/T]ATGTACAGTATATCAGAAGTCATCA
Phenotype
MIM: 600215
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MFAP1 PubMed Links

Gene Details

Gene
MFAP1
Gene Name
microfibrillar associated protein 1
There are no transcripts associated with this gene.

Gene
WDR76
Gene Name
WD repeat domain 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167941.1 Intron NP_001161413.1
NM_024908.3 Intron NP_079184.2
XM_005254680.3 Intron XP_005254737.1

View Full Product Details