Product Details

SNP ID
rs3088278
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:66499446 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCTCCTCTGTAGTAGGTTTCTT[C/T]TCTGCAGGCTTCTTTTCAGGGGCTG
Phenotype
MIM: 176872 MIM: 180479 MIM: 605979 MIM: 609984
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MAP2K1 PubMed Links

Gene Details

Gene
MAP2K1
Gene Name
mitogen-activated protein kinase kinase 1
There are no transcripts associated with this gene.

Gene
MIR4512
Gene Name
microRNA 4512
There are no transcripts associated with this gene.

Gene
RPL4
Gene Name
ribosomal protein L4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000968.3 Intron NP_000959.2
Gene
SNAPC5
Gene Name
small nuclear RNA activating complex polypeptide 5
There are no transcripts associated with this gene.

Gene
SNORD16
Gene Name
small nucleolar RNA, C/D box 16
There are no transcripts associated with this gene.

Gene
SNORD18A
Gene Name
small nucleolar RNA, C/D box 18A
There are no transcripts associated with this gene.

Gene
SNORD18B
Gene Name
small nucleolar RNA, C/D box 18B
There are no transcripts associated with this gene.

Gene
SNORD18C
Gene Name
small nucleolar RNA, C/D box 18C
There are no transcripts associated with this gene.

Gene
ZWILCH
Gene Name
zwilch kinetochore protein
There are no transcripts associated with this gene.

View Full Product Details