Product Details

SNP ID
rs34226451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74785561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAAATAACAGGATGGTAACACCT[A/G]CTTCAGAGGAAGGTGTAAGGACAGT
Phenotype
MIM: 124095
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSK PubMed Links

Gene Details

Gene
CSK
Gene Name
c-src tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127190.1 Intron NP_001120662.1
NM_004383.2 Intron NP_004374.1
XM_005254165.4 Intron XP_005254222.1
XM_017021925.1 Intron XP_016877414.1
Gene
MIR4513
Gene Name
microRNA 4513
There are no transcripts associated with this gene.

View Full Product Details