Product Details

SNP ID
rs28363828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75649522 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGTGGGCTGGGCACCAACGGGCA[C/T]CCTCCCCTCAACCTCTCCTACCCTG
Phenotype
MIM: 612980
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IMP3 PubMed Links

Gene Details

Gene
IMP3
Gene Name
IMP3, U3 small nucleolar ribonucleoprotein
There are no transcripts associated with this gene.

Gene
LOC107984799
Gene Name
translation initiation factor IF-2-like
There are no transcripts associated with this gene.

Gene
SNX33
Gene Name
sorting nexin 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318146.1 516 Silent Mutation CAC,CAT H140H NP_001305075.1
NM_153271.1 516 Silent Mutation CAC,CAT H140H NP_695003.1

View Full Product Details