Product Details

SNP ID
rs879091253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56608644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACCTGTCCCGACTCTAGCCGCCT[C/T]TTCAGCTCGCCATGGATCCCAACTG
Phenotype
MIM: 156358 MIM: 156360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MT1L PubMed Links

Gene Details

Gene
MT1L
Gene Name
metallothionein 1L (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
MT2A
Gene Name
metallothionein 2A
There are no transcripts associated with this gene.

View Full Product Details