Product Details

SNP ID
rs28524872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67192558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTACAGCTATGGGCCAGGCTCGGG[C/G]TGCAGGTGTTCGTCCTCGTGGCCCT
Phenotype
MIM: 600659 MIM: 606422 MIM: 614117 MIM: 613701
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
E2F4 PubMed Links

Gene Details

Gene
E2F4
Gene Name
E2F transcription factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001950.3 Intron NP_001941.2
Gene
ELMO3
Gene Name
engulfment and cell motility 3
There are no transcripts associated with this gene.

Gene
EXOC3L1
Gene Name
exocyst complex component 3 like 1
There are no transcripts associated with this gene.

Gene
KIAA0895L
Gene Name
KIAA0895 like
There are no transcripts associated with this gene.

Gene
MIR328
Gene Name
microRNA 328
There are no transcripts associated with this gene.

View Full Product Details