Product Details

SNP ID
hCV60401841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89561643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTGTGGATCCGAGGAGGCGGAA[C/T]AAGTCCACGGAGTCCCTGCAGGCCA
Phenotype
MIM: 113703 MIM: 602783
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPL13 PubMed Links

Gene Details

Gene
RPL13
Gene Name
ribosomal protein L13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000977.3 388 Silent Mutation AAC,AAT N104N NP_000968.2
NM_001243130.1 388 Silent Mutation AAC,AAT N85N NP_001230059.1
NM_001243131.1 388 Intron NP_001230060.1
NM_033251.2 388 Silent Mutation AAC,AAT N104N NP_150254.1
Gene
SNORD68
Gene Name
small nucleolar RNA, C/D box 68
There are no transcripts associated with this gene.

Gene
SPG7
Gene Name
SPG7, paraplegin matrix AAA peptidase subunit
There are no transcripts associated with this gene.

View Full Product Details