Product Details

SNP ID
rs34158934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89919542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGCGGGAGCAACGTGCTGGAGA[C/T]GGCCGTCATCCTCCTGCTGGAGGCC
Phenotype
MIM: 155555 MIM: 612326 MIM: 602661
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MC1R PubMed Links

Gene Details

Gene
MC1R
Gene Name
melanocortin 1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002386.3 1664 Missense Mutation ACG,ATG T95M NP_002377.4
Gene
TCF25
Gene Name
transcription factor 25
There are no transcripts associated with this gene.

Gene
TUBB3
Gene Name
tubulin beta 3 class III
There are no transcripts associated with this gene.

View Full Product Details