Product Details
- SNP ID
-
rs28479211
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:75913477 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGACTTTTTTTATTTCCTTTTTT[A/T]TTCCTGAGTCCAGAGTTACAAAAAG
- Phenotype
-
MIM: 616807
MIM: 611844
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FBF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201027437] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FBF1
- Gene Name
- Fas binding factor 1
- Gene
- MRPL38
- Gene Name
- mitochondrial ribosomal protein L38
There are no transcripts associated with this gene.
View Full Product Details