Product Details

SNP ID
rs34158267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:13883173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAAGCATTGCCTGTTCTGGAAGA[C/T]ACCATCCATCCTGCAGATCCTGAGC
Phenotype
MIM: 607397
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MC2R PubMed Links

Gene Details

Gene
MC2R
Gene Name
melanocortin 2 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000529.2 2552 UTR 3 NP_000520.1
NM_001291911.1 2552 UTR 3 NP_001278840.1
XM_017025781.1 2552 UTR 3 XP_016881270.1

View Full Product Details