Product Details

SNP ID
rs34448266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42402573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCCGGGTGCCCTGCTCTTCTCT[A/G]AATGCACCTGTACCGGCCCCCTCTG
Phenotype
MIM: 151750
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIPE PubMed Links

Gene Details

Gene
LIPE
Gene Name
lipase E, hormone sensitive type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005357.3 Intron NP_005348.2
XM_005258937.3 Intron XP_005258994.1
XM_005258938.4 Intron XP_005258995.1
XM_005258939.3 Intron XP_005258996.2
XM_005258940.3 Intron XP_005258997.1
XM_006723218.2 Intron XP_006723281.1
XM_017026810.1 Intron XP_016882299.1
Gene
LIPE-AS1
Gene Name
LIPE antisense RNA 1
There are no transcripts associated with this gene.

Gene
LOC101930071
Gene Name
uncharacterized LOC101930071
There are no transcripts associated with this gene.

View Full Product Details