Product Details

SNP ID
rs28450927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44171129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGTGTAGGATTTTTACTGTTTTA[A/G]GAACTACAGTGTTAGTTTTCTTGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF226 PubMed Links

Gene Details

Gene
ZNF226
Gene Name
zinc finger protein 226
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032372.1 Intron NP_001027544.1
NM_001032373.1 Intron NP_001027545.1
NM_001032374.1 Intron NP_001027546.1
NM_001146220.2 Intron NP_001139692.1
NM_001319088.1 Intron NP_001306017.1
NM_001319089.1 Intron NP_001306018.1
NM_001319090.1 Intron NP_001306019.1
NM_015919.3 Intron NP_057003.2
NM_016444.2 Intron NP_057528.2
XM_005259227.2 Intron XP_005259284.1
XM_006723367.3 Intron XP_006723430.1
XM_006723368.3 Intron XP_006723431.1
XM_006723369.2 Intron XP_006723432.1
XM_017027262.1 Intron XP_016882751.1
XM_017027263.1 Intron XP_016882752.1
XM_017027264.1 Intron XP_016882753.1
XM_017027265.1 Intron XP_016882754.1

View Full Product Details