Product Details

SNP ID
rs34339153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45005439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGAATACTCAGTGGCTCCAGTTA[G/T]CTGGGTCTGGGCCTTTCTCTGGAGA
Phenotype
MIM: 604758
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RELB PubMed Links

Gene Details

Gene
RELB
Gene Name
RELB proto-oncogene, NF-kB subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006509.3 Intron NP_006500.2
XM_005259127.3 Intron XP_005259184.1
XM_005259128.2 Intron XP_005259185.1

View Full Product Details