Product Details

SNP ID
rs28609887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46620763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGATGCCAGGGCTCCCAAGCCTG[A/G]TGGGGGACCAGCGGCAAGGGAAGGC
Phenotype
MIM: 114183 MIM: 600022
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CALM3 PubMed Links

Gene Details

Gene
CALM3
Gene Name
calmodulin 3
There are no transcripts associated with this gene.

Gene
PTGIR
Gene Name
prostaglandin I2 (prostacyclin) receptor (IP)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000960.3 2521 UTR 3 NP_000951.1
XM_005259093.2 2521 UTR 3 XP_005259150.1
XM_005259095.3 2521 UTR 3 XP_005259152.1

View Full Product Details