Product Details

SNP ID
rs28750161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52050674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGGCCTAAAAACTAGATCTAG[A/G]TAGCCAAAAAGATTACATTAAACTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF432 PubMed Links

Gene Details

Gene
ZNF432
Gene Name
zinc finger protein 432
There are no transcripts associated with this gene.

Gene
ZNF841
Gene Name
zinc finger protein 841
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136499.1 3523 Intron NP_001129971.1
NM_001321349.1 3523 Intron NP_001308278.1
XM_011526833.2 3523 Intron XP_011525135.1
XM_017026652.1 3523 Intron XP_016882141.1
XM_017026653.1 3523 Intron XP_016882142.1
XM_017026654.1 3523 Intron XP_016882143.1
XM_017026655.1 3523 Intron XP_016882144.1
XM_017026656.1 3523 Intron XP_016882145.1
XM_017026657.1 3523 Intron XP_016882146.1
XM_017026658.1 3523 Intron XP_016882147.1
XM_017026659.1 3523 UTR 3 XP_016882148.1
XM_017026660.1 3523 Intron XP_016882149.1
XM_017026661.1 3523 Intron XP_016882150.1
XM_017026662.1 3523 Intron XP_016882151.1
XM_017026663.1 3523 Intron XP_016882152.1

View Full Product Details