Product Details

SNP ID
rs28648863
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56810886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGGGTATGTATTATGCACACCA[A/G]TGGAGACACACATAATACACTGTTA
Phenotype
MIM: 601483
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PEG3 PubMed Links

Gene Details

Gene
PEG3
Gene Name
paternally expressed 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146184.1 7560 UTR 3 NP_001139656.1
NM_001146185.1 7560 UTR 3 NP_001139657.1
NM_001146186.1 7560 UTR 3 NP_001139658.1
NM_001146187.1 7560 UTR 3 NP_001139659.1
NM_006210.2 7560 UTR 3 NP_006201.1
Gene
PEG3-AS1
Gene Name
PEG3 antisense RNA 1
There are no transcripts associated with this gene.

Gene
ZIM2
Gene Name
zinc finger imprinted 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146326.1 7560 Intron NP_001139798.1
NM_001146327.1 7560 Intron NP_001139799.1
NM_015363.4 7560 Intron NP_056178.3

View Full Product Details