Product Details

SNP ID
rs35879596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35007986 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGCTCCGACAACTGAGTGGAAAGT[C/T]GCTGTTCAATAAATATTTGTTGTAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRAMD1A PubMed Links

Gene Details

Gene
GRAMD1A
Gene Name
GRAM domain containing 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136199.2 Intron NP_001129671.1
NM_001320034.1 Intron NP_001306963.1
NM_001320035.1 Intron NP_001306964.1
NM_001320036.1 Intron NP_001306965.1
NM_020895.4 Intron NP_065946.2
XM_011527149.1 Intron XP_011525451.1
XM_011527153.1 Intron XP_011525455.1
XM_011527154.2 Intron XP_011525456.1
XM_011527155.1 Intron XP_011525457.1
XM_011527156.1 Intron XP_011525458.1
XM_017027034.1 Intron XP_016882523.1
XM_017027035.1 Intron XP_016882524.1

View Full Product Details