Product Details

SNP ID
rs28469420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35125926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTCAGAACCCTCCCCTGCCTGCA[C/G]CTCACTCCAGAAACAGTGGGCAGTG
Phenotype
MIM: 604996 MIM: 608303
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FXYD3 PubMed Links

Gene Details

Gene
FXYD3
Gene Name
FXYD domain containing ion transport regulator 3
There are no transcripts associated with this gene.

Gene
LGI4
Gene Name
leucine rich repeat LGI family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139284.2 Intron NP_644813.1
XM_011526595.1 Intron XP_011524897.1
XM_017026428.1 Intron XP_016881917.1
XM_017026429.1 Intron XP_016881918.1
XM_017026430.1 Intron XP_016881919.1
Gene
MIR6887
Gene Name
microRNA 6887
There are no transcripts associated with this gene.

View Full Product Details