Product Details

SNP ID
rs11538357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18613106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCAGCTGCGGTGCCGCGACCGC[A/G]ACCTCGGCCCGCAGCCCTCGCAGGT
Phenotype
MIM: 604237
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRLF1 PubMed Links

Gene Details

Gene
CRLF1
Gene Name
cytokine receptor like factor 1
There are no transcripts associated with this gene.

Gene
TMEM59L
Gene Name
transmembrane protein 59 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012109.2 235 Missense Mutation AAC,GAC N50D NP_036241.1

View Full Product Details