Product Details

SNP ID
rs28372903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:16730079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGGCCTTAGGTGGGTTCGTGCGC[C/T]TTCTACCTCGCTGTTTCGGTTTTCC
Phenotype
MIM: 603520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNRPB2 PubMed Links

Gene Details

Gene
SNRPB2
Gene Name
small nuclear ribonucleoprotein polypeptide B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003092.4 116 UTR 5 NP_003083.1
NM_198220.2 116 Intron NP_937863.1

View Full Product Details