Product Details
- SNP ID
-
rs36059302
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:25563201 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATGAGGTCAAGTGGACATTATGT[G/T]TATATTCCAAAGCCAAAATTACTGC
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR155HG
PubMed Links
Gene Details
- Gene
- MIR155HG
- Gene Name
- MIR155 host gene
There are no transcripts associated with this gene.
View Full Product Details