Product Details

SNP ID
rs15682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:49962980 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGGTGGTGCCTGTCCCTGCGGCC[C/T]TCAGAGCGGCCGTCGCTGGATCAGA
Phenotype
MIM: 610580
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6821 PubMed Links

Gene Details

Gene
MIR6821
Gene Name
microRNA 6821
There are no transcripts associated with this gene.

Gene
PIM3
Gene Name
Pim-3 proto-oncogene, serine/threonine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001852.3 1287 Silent Mutation CCC,CCT P278P NP_001001852.2

View Full Product Details