Product Details

SNP ID
rs34723335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:102566886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTACAGGGTGTGGCAGCCCTCA[A/G]TGCTTCTCTTTCTCACACAGGCCTG
Phenotype
MIM: 300318
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NXF4 PubMed Links

Gene Details

Gene
NXF4
Gene Name
nuclear RNA export factor 4 pseudogene
There are no transcripts associated with this gene.

View Full Product Details