Product Details

SNP ID
rs28421327
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:140783851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCCAGTAACCTCCCAGTCTTCC[A/T]GAAAATCCACGTCTTCCCAACAATC
Phenotype
MIM: 302650
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CDR1 PubMed Links

Gene Details

Gene
CDR1
Gene Name
cerebellar degeneration related protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004065.2 708 Silent Mutation TCA,TCT S172S NP_004056.2

View Full Product Details