Product Details

SNP ID
rs28382703
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:123860881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGCCCTCTCCACCTTCGAAAAAG[A/G]AACAACGTCAATCCAACCTTTGTTA
Phenotype
MIM: 300079
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
XIAP PubMed Links

Gene Details

Gene
XIAP
Gene Name
X-linked inhibitor of apoptosis
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167.3 Intron NP_001158.2
NM_001204401.1 Intron NP_001191330.1
XM_006724754.2 Intron XP_006724817.1
XM_011531329.2 Intron XP_011529631.1

View Full Product Details