Product Details

SNP ID
rs34289137
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:19589563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATGCAAGCCCTAAACACTCTTT[C/T]GACTCCAGAGGAGAAGCTGGCAGCT
Phenotype
MIM: 400031
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TXLNGY PubMed Links

Gene Details

Gene
TXLNGY
Gene Name
taxilin gamma pseudogene, Y-linked
There are no transcripts associated with this gene.

View Full Product Details