Product Details

SNP ID
rs34194308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:20766458 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATCAAGGACAGGATTTCTTAAAT[C/G]TAGCAGGGAGTAGAGCTGGAGAGAT
Phenotype
MIM: 400030
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RPS4Y2 PubMed Links

Gene Details

Gene
RPS4Y2
Gene Name
ribosomal protein S4, Y-linked 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039567.2 Intron NP_001034656.1

View Full Product Details