Product Details
- SNP ID
-
rs28604487
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:94765167 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATCTTGGCAGGAAAATTAGAAATT[C/T]CATATAAACCTTAAGAAAATAAAGT
- Phenotype
-
MIM: 603248
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BMPR1B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114682420] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BMPR1B
- Gene Name
- bone morphogenetic protein receptor type 1B
- Gene
- BMPR1B-AS1
- Gene Name
- BMPR1B antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details