Product Details
- SNP ID
-
rs11554389
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:99949687 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTGCAAGCCGGCTCTCTGCGAGCG[G/T]GAAACCGCCTTTGTCTTGGCCTTTC
- Phenotype
-
MIM: 142763
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DNAJB14
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79002079] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DNAJB14
- Gene Name
- DnaJ heat shock protein family (Hsp40) member B14
There are no transcripts associated with this gene.
- Gene
- H2AFZ
- Gene Name
- H2A histone family member Z
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002106.3 |
|
Intron |
|
|
NP_002097.1 |
- Gene
- LOC256880
- Gene Name
- uncharacterized LOC256880
There are no transcripts associated with this gene.
View Full Product Details