Product Details

SNP ID
rs1143561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:108622007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGTAGGCTTTCCTACAATACAGC[C/G]TCTAACAAAACTAGGCTGTAAGTAT
Phenotype
MIM: 616862
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RPL34 PubMed Links

Gene Details

Gene
RPL34
Gene Name
ribosomal protein L34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000995.4 100 Silent Mutation GCC,GCG A16A NP_000986.2
NM_001319232.1 100 Intron NP_001306161.1
NM_001319234.1 100 Silent Mutation GCC,GCG A16A NP_001306163.1
NM_001319235.1 100 Intron NP_001306164.1
NM_001319236.1 100 Intron NP_001306165.1
NM_033625.3 100 Silent Mutation GCC,GCG A16A NP_296374.1
Gene
RPL34-AS1
Gene Name
RPL34 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details