Product Details

SNP ID
rs28716085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:152538657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATACATAAAGCAATTATATAGT[A/G]TAAGTGATTATAAAGGAAACCACCA
Phenotype
MIM: 606278
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DKFZP434I0714 PubMed Links

Gene Details

Gene
DKFZP434I0714
Gene Name
uncharacterized protein DKFZP434I0714
There are no transcripts associated with this gene.

Gene
FBXW7
Gene Name
F-box and WD repeat domain containing 7
There are no transcripts associated with this gene.

Gene
MIR4453
Gene Name
microRNA 4453
There are no transcripts associated with this gene.

View Full Product Details