Product Details
- SNP ID
-
rs28395809
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:48431231 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAAGTTATGGACTTGGAGGAGAGA[A/G]CCAAGAATGGAGTTATACGGGGGCA
- Phenotype
-
MIM: 610585
MIM: 601158
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGAP22
PubMed Links
Gene Details
- Gene
- ARHGAP22
- Gene Name
- Rho GTPase activating protein 22
- Gene
- MAPK8
- Gene Name
- mitogen-activated protein kinase 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001278547.1 |
1331 |
Missense Mutation |
ACC,GCC |
T367A |
NP_001265476.1 |
NM_001278548.1 |
1331 |
Missense Mutation |
ACC,GCC |
T291A |
NP_001265477.1 |
NM_001323302.1 |
1331 |
Missense Mutation |
ACC,GCC |
T367A |
NP_001310231.1 |
NM_001323320.1 |
1331 |
Missense Mutation |
ACC,GCC |
T291A |
NP_001310249.1 |
NM_001323321.1 |
1331 |
Missense Mutation |
ACC,GCC |
T367A |
NP_001310250.1 |
NM_001323322.1 |
1331 |
Missense Mutation |
ACC,GCC |
T367A |
NP_001310251.1 |
NM_001323323.1 |
1331 |
Missense Mutation |
ACC,GCC |
T367A |
NP_001310252.1 |
NM_001323324.1 |
1331 |
Missense Mutation |
ACC,GCC |
T367A |
NP_001310253.1 |
NM_001323325.1 |
1331 |
Missense Mutation |
ACC,GCC |
T367A |
NP_001310254.1 |
NM_001323326.1 |
1331 |
Missense Mutation |
ACC,GCC |
T367A |
NP_001310255.1 |
NM_001323327.1 |
1331 |
Missense Mutation |
ACC,GCC |
T367A |
NP_001310256.1 |
NM_001323328.1 |
1331 |
Missense Mutation |
ACC,GCC |
T367A |
NP_001310257.1 |
NM_001323329.1 |
1331 |
Missense Mutation |
ACC,GCC |
T367A |
NP_001310258.1 |
NM_001323330.1 |
1331 |
Missense Mutation |
ACC,GCC |
T367A |
NP_001310259.1 |
NM_001323331.1 |
1331 |
Missense Mutation |
ACC,GCC |
T367A |
NP_001310260.1 |
NM_139046.3 |
1331 |
Missense Mutation |
ACC,GCC |
T367A |
NP_620634.1 |
NM_139049.3 |
1331 |
Missense Mutation |
ACC,GCC |
T367A |
NP_620637.1 |
View Full Product Details