Product Details

SNP ID
rs35575576
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:117150472 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAACATTCTAGTACACTTCCTACA[C/T]TTCCCTTTCAGACTAGATTTATTTA
Phenotype
MIM: 608162
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VTCN1 PubMed Links

Gene Details

Gene
VTCN1
Gene Name
V-set domain containing T cell activation inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253849.1 Intron NP_001240778.1
NM_001253850.1 Intron NP_001240779.1
NM_024626.3 Intron NP_078902.2
XM_011542143.2 Intron XP_011540445.2
XM_017002335.1 Intron XP_016857824.1

View Full Product Details