Product Details
- SNP ID
-
hCV61863779
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:87783491 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTGCTCATTGCCTCCTTGTATTTC[A/T]GAATGAGTTTCTGCTTCTCTTCTTT
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MKRN9P
PubMed Links
Gene Details
- Gene
- MKRN9P
- Gene Name
- makorin ring finger protein 9, pseudogene
There are no transcripts associated with this gene.
View Full Product Details