Product Details

SNP ID
rs669174
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:222525207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGGGAGTTAAATTACCTGCCAA[A/C]TGAATCAGAAAACTGCCAAAGAAAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HHIPL2 PubMed Links

Gene Details

Gene
HHIPL2
Gene Name
HHIP like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024746.3 1821 Intron NP_079022.2
XM_011509985.2 1821 Intron XP_011508287.1
XM_011509986.1 1821 Missense Mutation CAG,CAT Q606H XP_011508288.1
XM_017002350.1 1821 Intron XP_016857839.1

View Full Product Details