Product Details

SNP ID
rs34477566
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:10002348 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTACTCGAATGTCTTATTTTCT[A/T]TTTAACATAATCTAGTAGATGAAAA
Phenotype
MIM: 608700 MIM: 608604
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NMNAT1 PubMed Links

Gene Details

Gene
NMNAT1
Gene Name
nicotinamide nucleotide adenylyltransferase 1
There are no transcripts associated with this gene.

Gene
RBP7
Gene Name
retinol binding protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052960.2 Intron NP_443192.1

View Full Product Details