Product Details

SNP ID
rs11551462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:47160802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTTGCTGTCATCATTTGTACAA[G/A]CTCTGAAAAAGAAGAAGTACACAAA
Phenotype
MIM: 114182
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
C2orf61 PubMed Links

Gene Details

Gene
C2orf61
Gene Name
chromosome 2 open reading frame 61
There are no transcripts associated with this gene.

Gene
CALM2
Gene Name
calmodulin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305624.1 650 Missense Mutation CTT,TTT L190F NP_001292553.1
NM_001305625.1 650 Missense Mutation CTT,TTT L106F NP_001292554.1
NM_001305626.1 650 Missense Mutation CTT,TTT L106F NP_001292555.1
NM_001743.5 650 Missense Mutation CTT,TTT L142F NP_001734.1

View Full Product Details