Product Details

SNP ID
rs17850955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73640949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAATACACACAGAAAGAGATCAC[A/T]GACTCCCTACCTTAGAAGAAGGGAG
Phenotype
MIM: 606716
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ALMS1P1 PubMed Links

Gene Details

Gene
ALMS1P1
Gene Name
ALMS1, centrosome and basal body associated protein pseudogene 1
There are no transcripts associated with this gene.

Gene
NAT8
Gene Name
N-acetyltransferase 8 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003960.3 847 Missense Mutation CAG,CTG Q227L NP_003951.3

View Full Product Details