Product Details

SNP ID
rs17876035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177402327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGCTCCCGGATCCAGGCCAGGT[A/G]GTAGGCCACATCGGTGTAGACGCCT
Phenotype
MIM: 610619 MIM: 612812 MIM: 182309
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
F12 PubMed Links

Gene Details

Gene
F12
Gene Name
coagulation factor XII
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000505.3 1862 Missense Mutation CAC,TAC H605Y NP_000496.2
XM_011534462.2 1862 Missense Mutation CAC,TAC H493Y XP_011532764.1
Gene
PFN3
Gene Name
profilin 3
There are no transcripts associated with this gene.

Gene
SLC34A1
Gene Name
solute carrier family 34 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167579.1 1862 Intron NP_001161051.1
NM_003052.4 1862 Intron NP_003043.3
XM_005265975.1 1862 Intron XP_005266032.1
XM_017009773.1 1862 Intron XP_016865262.1
XM_017009774.1 1862 Intron XP_016865263.1
XM_017009775.1 1862 Intron XP_016865264.1

View Full Product Details