Product Details
- SNP ID
-
rs4990389
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:179644891 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCAGGATCACCAGGCTGAACACAA[T/C]AAAGAGGACGAAATGGATGATCTGG
- Phenotype
-
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C5orf60
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4990390] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C5orf60
- Gene Name
- chromosome 5 open reading frame 60
View Full Product Details