Product Details

SNP ID
rs28480047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:23843072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCACCAATAGTTTCTACCTAAG[C/T]GAGTTTGGAGTGGCCCCTGCAGTCC
Phenotype
MIM: 601185
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STC1 PubMed Links

Gene Details

Gene
STC1
Gene Name
stanniocalcin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003155.2 2726 UTR 3 NP_003146.1

View Full Product Details