Product Details
- SNP ID
-
rs34418873
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:136980863 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCATGACGGAACAATAGGTGAGCC[A/T]CTCCATTCATTCATTCATTCATTCA
- Phenotype
-
MIM: 176803
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LCNL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs571822377] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LCNL1
- Gene Name
- lipocalin like 1
There are no transcripts associated with this gene.
- Gene
- PRR31
- Gene Name
- proline rich 31
There are no transcripts associated with this gene.
- Gene
- PTGDS
- Gene Name
- prostaglandin D2 synthase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000954.5 |
|
Intron |
|
|
NP_000945.3 |
View Full Product Details