Product Details

SNP ID
rs28687338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136329288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGCCCTCCAGCAGGGCCTGTGA[C/G]GAGGTCCCCCCGGGAGTCTGCACCC
Phenotype
MIM: 609491
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DKFZP434A062 PubMed Links

Gene Details

Gene
DKFZP434A062
Gene Name
uncharacterized LOC26102
There are no transcripts associated with this gene.

Gene
GPSM1
Gene Name
G-protein signaling modulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145638.2 Intron NP_001139110.2
NM_001145639.1 Intron NP_001139111.1
NM_001200003.1 Intron NP_001186932.1
NM_015597.5 Intron NP_056412.5
XM_011518498.2 Intron XP_011516800.1
XM_017014598.1 Intron XP_016870087.1
XM_017014599.1 Intron XP_016870088.1

View Full Product Details