Product Details

SNP ID
rs28710427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:46126186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGCATTTCAATTGCTTTGAAAA[A/G]TGGCTAAGTTCCATTACATACCACC
Phenotype
MIM: 153430
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LCP1 PubMed Links

Gene Details

Gene
LCP1
Gene Name
lymphocyte cytosolic protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002298.4 3425 UTR 3 NP_002289.2
XM_005266374.1 3425 UTR 3 XP_005266431.1

View Full Product Details