Product Details
- SNP ID
-
hCV63082627
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:131576682 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTCAGCCACGAGGAGGGGTGGAAG[A/G]AGGCCCTACAGTATTGAGAAAGGCT
- Phenotype
-
MIM: 608313
MIM: 605042
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARG1
PubMed Links
Gene Details
- Gene
- ARG1
- Gene Name
- arginase 1
There are no transcripts associated with this gene.
- Gene
- MED23
- Gene Name
- mediator complex subunit 23
There are no transcripts associated with this gene.
View Full Product Details