Product Details
- SNP ID
-
rs34815843
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:132744533 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAGTACAGTCAGCTTTCTTAACA[C/T]AGAAGTTAAAAGCAAATTAATCATT
- Phenotype
-
MIM: 603571
MIM: 606592
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
VNN2
PubMed Links
Gene Details
- Gene
- VNN2
- Gene Name
- vanin 2
- Gene
- VNN3
- Gene Name
- vanin 3
There are no transcripts associated with this gene.
View Full Product Details