Product Details

SNP ID
rs28362173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:137147796 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATCTTCTATAGATACATTTTTTT[C/T]CTTTTGGTATCTATATGGTAAATTT
Phenotype
MIM: 606648
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL22RA2 PubMed Links

Gene Details

Gene
IL22RA2
Gene Name
interleukin 22 receptor subunit alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052962.2 773 Missense Mutation AAA,GAA K190E NP_443194.1
NM_181309.1 773 Missense Mutation AAA,GAA K158E NP_851826.1
NM_181310.1 773 Intron NP_851827.1

View Full Product Details