Product Details

SNP ID
rs17882836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20470880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGATTGAGCCTATAAGATAGAT[A/G]GGGCCAATTTACATCCCTTGTTTAT
Phenotype
MIM: 164050 MIM: 609865
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PNP PubMed Links

Gene Details

Gene
PNP
Gene Name
purine nucleoside phosphorylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000270.3 Intron NP_000261.2
Gene
TMEM55B
Gene Name
transmembrane protein 55B
There are no transcripts associated with this gene.

View Full Product Details